Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025624.2 | NP_001020795.1 | 6 | 1038 | ||
GenBank mRNA | BC099178.1 | - | 6 | 1005 | 7383003 | |
BC099815.1 | - | 6 | 1013 | 7929654 | ||
FQ213300.1 | - | 6 | 1005 | |||
FQ213727.1 | - | 6 | 998 | |||
FQ214234.1 | - | 4 | 831 | |||
FQ214394.1 | - | 6 | 997 | |||
FQ214422.1 | - | 6 | 1008 | |||
FQ219745.1 | - | 6 | 1073 | |||
FQ219836.1 | - | 6 | 1035 | |||
FQ220601.1 | - | 6 | 1049 | |||
FQ221300.1 | - | 6 | 979 | |||
FQ223249.1 | - | 6 | 996 |