Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025624.2 | NP_001020795.1 | 6 | 622 | ||
GenBank mRNA | AF329828.1 | - | 1 | 158 | ||
BC099178.1 | - | 6 | 588 | 7383003 | ||
BC099815.1 | - | 6 | 597 | 7929654 | ||
FQ213300.1 | - | 6 | 589 | |||
FQ213727.1 | - | 6 | 582 | |||
FQ214234.1 | - | 4 | 415 | |||
FQ214394.1 | - | 6 | 581 | |||
FQ214422.1 | - | 6 | 592 | |||
FQ215102.1 | - | 6 | 599 | |||
FQ216264.1 | - | 6 | 615 | |||
FQ216489.1 | - | 6 | 554 | |||
FQ217912.1 | - | 6 | 591 | |||
FQ219745.1 | - | 6 | 657 | |||
FQ220601.1 | - | 6 | 633 | |||
FQ221300.1 | - | 6 | 563 | |||
FQ223249.1 | - | 6 | 580 | |||
FQ229179.1 | - | 6 | 581 | |||
FQ229184.1 | - | 6 | 554 |
Gene Symbol: LOC100360316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751587.3 | XP_003751635.1 | 1 | 585 |