Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128427.2 | NP_001121899.1 | 2 | 523 | ||
NM_001128428.2 | NP_001121900.1 | 2 | 523 | |||
NM_020116.4 | NP_064501.2 | 2 | 523 | |||
XM_011532126.1 | XP_011530428.1 | 2 | 489 | |||
GenBank mRNA | AB033089.1 | - | 2 | 453 | ||
AK315152.1 | - | 2 | 494 | |||
BC036502.2 | - | 2 | 524 | 5265170 | ||
BC073978.1 | - | 2 | 226 | |||
BI549596.1 | - | 534 | ||||
DA243182.1 | - | 2 | 506 |