Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025631.1 | NP_001020802.1 | 2 | 1062 | ||
GenBank mRNA | BC097476.1 | - | 2 | 1062 | 7457927 |