Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ceacam11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025404.1 | NP_001020575.1 | 2 | 243 | ||
GenBank mRNA | BC097990.1 | - | 2 | 243 | 7445748 | |
U66293.1 | - | 2 | 285 |