Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001313941.1 | NP_001300870.1 | 3, 4 | 524 | ||
NM_001313943.1 | NP_001300872.1 | 2, 3 | 429 | |||
NM_001313945.1 | NP_001300874.1 | 2, 3 | 306 | |||
NM_001664.3 | NP_001655.1 | 2, 3 | 429 | |||
GenBank mRNA | AF498970.1 | - | 1, 2 | 149 | ||
AK130066.1 | - | 2, 3 | 425 | |||
AK130808.1 | - | 2, 3 | 305 | |||
AK222556.1 | - | 2, 3 | 321 | |||
AK307220.1 | - | 2, 3 | 204 | |||
BC001360.2 | - | 2, 3 | 261 | 3051460 | ||
BC005976.1 | - | 2, 3 | 266 | 4102976 | ||
BF966305.1 | - | 3, 4 | 399 | |||
BQ721697.1 | - | 2, 3 | 302 | |||
BT019870.1 | - | 1, 2 | 149 | |||
BX647063.1 | - | 2, 3 | 325 | |||
L09159.1 | - | 1, 2 | 138 | |||
L25080.1 | - | 2, 3 | 300 | |||
X05026.1 | - | 2, 3 | 307 |