Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025643.1 | NP_001020814.1 | 3 | 427 | ||
GenBank mRNA | BC099198.1 | - | 3 | 427 | 7383920 |