Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025646.1 | NP_001020817.1 | 5 | 718 | ||
XM_006231318.2 | XP_006231380.1 | 5 | 698 | |||
XM_006231319.2 | XP_006231381.1 | 5 | 812 | |||
XM_006231320.2 | XP_006231382.1 | 5 | 655 | |||
GenBank mRNA | BC098013.1 | - | 5 | 718 | 7454372 |