Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126337.1 | NP_001119809.1 | 2 | 138 | ||
NM_001301317.1 | NP_001288246.1 | 4 | 348 | |||
NM_020127.2 | NP_064512.1 | 3 | 213 | |||
XM_017002223.1 | XP_016857712.1 | 3 | 289 | |||
XM_017002224.1 | XP_016857713.1 | 2 | 410 | |||
GenBank mRNA | AF254260.1 | - | 3 | 213 | ||
AK075001.1 | - | 2 | 135 | |||
AK223161.1 | - | 3 | 209 | |||
AK298066.1 | - | 4 | 348 | |||
AK299263.1 | - | 2 | 136 | |||
AK315414.1 | - | 3 | 209 | |||
AL136917.1 | - | 3 | 201 | |||
AY358610.1 | - | 2 | 132 | |||
BC002933.1 | - | 2 | 106 | 3956873 | ||
BC008301.1 | - | 3 | 193 | 3506004 |