Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025647.1 | NP_001020818.1 | 7, 8 | 1023 | ||
XM_006231821.1 | XP_006231883.1 | 5, 6 | 719 | |||
XM_008760664.1 | XP_008758886.1 | 4, 5 | 764 | |||
XM_008760665.1 | XP_008758887.1 | 5, 6 | 642 | |||
GenBank mRNA | BC097489.1 | - | 7, 8 | 1023 | 7460568 |