Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025410.1 | NP_001020581.1 | 8 | 1088 | ||
GenBank mRNA | BC086333.1 | - | 7 | 950 | ||
BC097963.1 | - | 8 | 1088 | 7387550 | ||
FQ214132.1 | - | 8 | 1116 |