Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025664.1 | NP_001020835.2 | 2, 3 | 677 | ||
NM_001042561.1 | NP_001036026.1 | 7, 8 | 1089 | |||
XM_008768016.1 | XP_008766238.1 | 7, 8 | 2012 | |||
GenBank mRNA | BC098720.1 | - | 2, 3 | 677 | ||
BC167746.1 | - | 7, 8 | 1092 | 8364583 | ||
CK473503.1 | - | 5, 6 | 572 |