Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025664.1 | NP_001020835.2 | 4 | 1309 | ||
NM_001042561.1 | NP_001036026.1 | 9 | 1721 | |||
XM_008768016.1 | XP_008766238.1 | 9 | 2644 | |||
GenBank mRNA | BC098720.1 | - | 4 | 1309 | ||
BC167746.1 | - | 9 | 1724 | 8364583 |