Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322248.1 | NP_001309177.1 | 3 | 764 | ||
NM_020141.3 | NP_064526.1 | 3 | 663 | |||
NR_136242.1 | 4 | 792 | ||||
NR_136243.1 | 2 | 571 | ||||
GenBank mRNA | AF164793.1 | - | 3 | 684 | ||
AK075191.1 | - | 3 | 685 | |||
AK310192.1 | - | 2 | 551 | |||
BC008739.2 | - | 3 | 663 | 3346340 | ||
BC065186.1 | - | 3 | 663 | 4642864 | ||
BC066918.1 | - | 3 | 692 | 4826492 | ||
BC067116.1 | - | 3 | 650 | 5532892 | ||
BC171849.1 | - | 3 | 1641 | 9054503 | ||
BG724370.1 | - | 2 | 576 | |||
BX331284.2 | - | 4 | 753 | |||
DA782106.1 | - | 2 | 539 |