Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025665.1 | NP_001020836.1 | 6 | 817 | ||
GenBank mRNA | BC097409.1 | - | 6 | 817 | 7454160 |