Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025679.1 | NP_001020850.1 | 5 | 1366 | ||
XM_006228417.1 | XP_006228479.1 | 5 | 1099 | |||
GenBank mRNA | BC097499.1 | - | 5 | 1366 | 7462593 |