Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025679.1 | NP_001020850.1 | 4 | 991 | ||
GenBank mRNA | BC097499.1 | - | 4 | 991 | 7462593 |