Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ch25h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025415.1 | NP_001020586.1 | 1 | 980 | ||
XM_006231291.2 | XP_006231353.1 | 2 | 3581 | |||
GenBank mRNA | BC097964.1 | - | 1 | 980 | 7387594 |