Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020151.3 | NP_064536.2 | 4 | 1009 | ||
XM_011511499.1 | XP_011509801.1 | 4 | 626 | |||
GenBank mRNA | AF270647.1 | - | 4 | 936 | ||
AK054615.1 | - | 4 | 375 | |||
AK291190.1 | - | 4 | 391 | |||
BC003028.1 | - | 4 | 992 | |||
BC008894.2 | - | 4 | 992 | 3833465 | ||
BC012774.2 | - | 4 | 959 | 3842611 | ||
BC012793.2 | - | 4 | 973 | 4080200 | ||
BC013279.2 | - | 4 | 992 | 3830320 | ||
BC014274.2 | - | 4 | 988 | 3677198 | ||
BC032106.1 | - | 4 | 983 | 4643387 |