Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025689.1 | NP_001020860.1 | 6 | 566 | ||
GenBank mRNA | BC097427.1 | - | 6 | 566 | ||
BC158645.1 | - | 6 | 514 | 8363877 | ||
FQ228714.1 | - | 6 | 542 | |||
FQ228827.1 | - | 6 | 542 |