Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K7CL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286617.1 | NP_001273546.1 | 5 | 581 | ||
NM_001286618.1 | NP_001273547.1 | 5 | 581 | |||
NM_001286619.1 | NP_001273548.1 | 1 | 248 | |||
NM_001286622.1 | NP_001273551.1 | 5 | 581 | |||
NM_001286623.1 | NP_001273552.1 | 6 | 738 | |||
NM_001286634.1 | NP_001273563.1 | 5 | 581 | |||
NM_020152.3 | NP_064537.1 | 7 | 810 | |||
GenBank mRNA | AF269161.1 | - | 5 | 419 | ||
AF269162.1 | - | 5 | 419 | |||
AF269163.1 | - | 5 | 419 | |||
AK122780.1 | - | 1 | 248 | |||
AK294287.1 | - | 5 | 561 | |||
AY171599.2 | - | 6 | 567 | |||
EF219157.1 | - | 6 | 738 |