Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025692.1 | NP_001020863.1 | 8, 9 | 906 | ||
XM_008762325.1 | XP_008760547.1 | 6, 7 | 2882 | |||
XM_008762326.1 | XP_008760548.1 | 9, 10 | 1887 | |||
GenBank mRNA | BC099095.1 | - | 8, 9 | 906 | 7459023 |