Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdcbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025692.1 | NP_001020863.1 | 7, 8 | 828 | ||
XM_008762325.1 | XP_008760547.1 | 5, 6 | 2804 | |||
XM_008762326.1 | XP_008760548.1 | 8, 9 | 1809 | |||
GenBank mRNA | BC099095.1 | - | 7, 8 | 828 | 7459023 |