Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025418.1 | NP_001020589.1 | 13 | 1577 | ||
XM_006243032.2 | XP_006243094.1 | 15 | 6687 | |||
XM_006243033.2 | XP_006243095.1 | 14 | 6244 | |||
XM_006243034.2 | XP_006243096.1 | 15 | 6692 | |||
XM_008766260.1 | XP_008764482.1 | 16 | 5305 | |||
XM_008766261.1 | XP_008764483.1 | 16 | 5672 | |||
GenBank mRNA | BC098007.1 | - | 13 | 1577 | 7453957 |