Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pxdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025719.1 | NP_001020890.1 | 3 | 529 | ||
XM_006253841.2 | XP_006253903.1 | 3 | 1181 | |||
GenBank mRNA | BC099145.1 | - | 3 | 529 | 7325668 |