Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020165.3 | NP_064550.3 | 7 | 903 | ||
XM_017006873.1 | XP_016862362.1 | 8 | 1061 | |||
GenBank mRNA | AB035274.2 | - | 7 | 938 | ||
AF169796.1 | - | 7 | 883 | |||
AK023075.1 | - | 7 | 890 | |||
AK222771.1 | - | 7 | 881 | |||
AY004333.1 | - | 7 | 849 | |||
BC001302.1 | - | 7 | 870 | 3451960 |