Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025730.2 | NP_001020901.1 | 9 | 1734 | ||
XM_006233623.2 | XP_006233685.1 | 8 | 2093 | |||
XM_006233624.2 | XP_006233686.1 | 10 | 1796 | |||
XM_006233625.2 | XP_006233687.1 | 10 | 2038 | |||
XM_006233626.2 | XP_006233688.1 | 9 | 1690 | |||
XM_006233627.2 | XP_006233689.1 | 9 | 1934 | |||
XM_006233628.2 | XP_006233690.1 | 8 | 1450 | |||
GenBank mRNA | BC099179.1 | - | 9 | 1737 | 7383052 |