Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077238.1 | NP_001070706.1 | 13 | 1389 | ||
NM_152988.2 | NP_694533.1 | 13 | 1389 | |||
XM_011528138.2 | XP_011526440.1 | 13 | 1445 | |||
XM_011528139.1 | XP_011526441.1 | 12 | 1247 | |||
XM_011528140.1 | XP_011526442.1 | 9 | 800 | |||
XM_017027017.1 | XP_016882506.1 | 14 | 1583 | |||
XM_017027018.1 | XP_016882507.1 | 12 | 1244 | |||
XM_017027019.1 | XP_016882508.1 | 14 | 1576 | |||
GenBank mRNA | AB040965.1 | - | 13 | 1338 | ||
AJ345027.1 | - | 13 | 1342 | |||
AJ420897.1 | - | 13 | 1338 | |||
AY169315.1 | - | 13 | 1337 | |||
BC028391.2 | - | 13 | 1326 | 4822524 | ||
BC093046.1 | - | 13 | 1389 | 30395602 |