Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025736.1 | NP_001020907.1 | 11 | 1266 | ||
GenBank mRNA | BC099173.1 | - | 11 | 1266 | 7382617 |