Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smpdl3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025737.1 | NP_001020908.1 | 3, 4 | 479 | ||
GenBank mRNA | BC097983.1 | - | 3, 4 | 479 | 7443444 |