Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025738.1 | NP_001020909.1 | 2 | 247 | ||
XM_006239220.2 | XP_006239282.1 | 2 | 290 | |||
XM_006239221.2 | XP_006239283.1 | 2 | 290 | |||
XM_006239222.2 | XP_006239284.1 | 2 | 288 | |||
XM_006239223.2 | XP_006239285.1 | 2 | 289 | |||
GenBank mRNA | BC098831.1 | - | 2 | 247 | 7378554 | |
FQ227288.1 | - | 2 | 268 |