Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dazap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025742.1 | NP_001020913.1 | 7 | 667 | ||
XM_006240968.2 | XP_006241030.1 | 7 | 734 | |||
XM_006240969.2 | XP_006241031.1 | 7 | 616 | |||
XM_006240970.2 | XP_006241032.1 | 7 | 613 | |||
XM_008765123.1 | XP_008763345.1 | 7 | 1242 | |||
GenBank mRNA | BC098930.1 | - | 7 | 667 | 7133199 |