Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025424.1 | NP_001020595.1 | 9 | 977 | ||
GenBank mRNA | BC129075.1 | - | 9 | 951 | 7383067 | |
DQ076481.1 | - | 9 | 977 |