Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025749.1 | NP_001020920.1 | 5 | 1504 | ||
GenBank mRNA | BC098740.1 | - | 5 | 1504 | 7386199 | |
FQ228943.1 | - | 5 | 1472 |