Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025766.1 | NP_001020937.1 | 2, 3 | 534 | ||
XM_006255599.2 | XP_006255661.1 | 3, 4 | 897 | |||
XM_006255600.2 | XP_006255662.1 | 3, 4 | 897 | |||
XM_008772527.1 | XP_008770749.1 | 3, 4 | 602 | |||
GenBank mRNA | BC076391.1 | - | 3, 4 | 660 | ||
BC098808.1 | - | 2, 3 | 534 | 7376793 |