Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC499742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024308.1 | NP_001019479.1 | 3 | 398 | ||
GenBank mRNA | AY539888.1 | - | 3 | 398 |