Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031068.1 | NP_112330.1 | 12 | 2059 | ||
GenBank mRNA | BC098014.1 | - | 12 | 2059 | 7454624 | |
FQ212256.1 | - | 12 | 2261 |