Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031068.1 | NP_112330.1 | 1, 2 | 134 | ||
GenBank mRNA | AB292042.1 | - | 1, 2 | 58 | ||
AB292043.1 | - | 1, 2 | 47 | |||
BC098014.1 | - | 1, 2 | 134 | 7454624 | ||
FQ212256.1 | - | 1, 2 | 336 |