Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030021.1 | NP_001025192.1 | 2 | 108 | ||
GenBank mRNA | BC099697.1 | - | 2 | 108 | 7934014 | |
FQ217138.1 | - | 2 | 121 | |||
FQ217658.1 | - | 2 | 122 | |||
FQ221181.1 | - | 2 | 121 | |||
FQ221584.1 | - | 2 | 120 | |||
FQ221984.1 | - | 2 | 122 | |||
FQ222461.1 | - | 2 | 120 | |||
FQ222498.1 | - | 2 | 121 | |||
FQ223422.1 | - | 2 | 123 | |||
FQ228678.1 | - | 2 | 122 | |||
X15098.1 | - | 2 | 122 |
Gene Symbol: LOC100911575 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749013.2 | XP_003749061.1 | 2 | 343 |