Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033655.2 | NP_001028827.2 | 8 | 1255 | ||
XM_008770977.1 | XP_008769199.1 | 8 | 1313 | |||
XM_008770978.1 | XP_008769200.1 | 8 | 1313 | |||
XM_008770979.1 | XP_008769201.1 | 8 | 1313 |