Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034848.2 | NP_001030020.2 | 4 | 465 | ||
GenBank mRNA | BC166872.1 | - | 4 | 465 | 6922077 | |
BC168778.1 | - | 519 | ||||
CB316202.1 | - | 4 | 468 | |||
U62779.1 | - | 4 | 464 | |||
V01237.1 | - | 4 | 441 |