Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031638.1 | NP_001026808.1 | 3 | 569 | ||
GenBank mRNA | BC098931.1 | - | 3 | 569 | 7368498 |