Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034126.1 | NP_001029298.1 | 10 | 876 | ||
GenBank mRNA | BC082034.1 | - | 9 | 1898 | ||
BC100637.1 | - | 10 | 876 | 5621093 | ||
FQ219054.1 | - | 7 | 808 | |||
FQ219183.1 | - | 10 | 882 | |||
FQ229934.1 | - | 10 | 889 |