Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031640.1 | NP_001026810.1 | 8 | 1310 | ||
GenBank mRNA | BC098728.1 | - | 8 | 1310 | 7381504 |