Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sap18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033685.2 | NP_001028857.2 | 4 | 562 | ||
GenBank mRNA | AW888346.1 | - | 1 | 230 | ||
BC104706.1 | - | 4 | 551 | 7316605 | ||
CK357541.1 | - | 4 | 519 | |||
FQ211269.1 | - | 3 | 411 | |||
FQ212380.1 | - | 4 | 534 | |||
FQ223271.1 | - | 4 | 529 | |||
FQ234134.1 | - | 4 | 570 |
Gene Symbol: RGD1561590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002727811.4 | XP_002727857.1 | 1 | 585 |