Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030026.1 | NP_001025197.1 | 7 | 870 | ||
GenBank mRNA | BC088256.1 | - | 7 | 734 | ||
BC099774.1 | - | 7 | 870 | 7932222 | ||
FQ232607.1 | - | 7 | 756 | |||
FQ233058.1 | - | 7 | 756 | |||
FQ233214.1 | - | 7 | 756 |