Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRGC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019612.3 | NP_062558.1 | 2 | 935 | ||
XM_005259058.3 | XP_005259115.1 | 2 | 1029 | |||
XM_006723283.3 | XP_006723346.1 | 2 | 966 | |||
XM_011527118.2 | XP_011525420.1 | 3 | 969 | |||
GenBank mRNA | BC033853.1 | - | 2 | 935 | ||
BC066939.1 | - | 2 | 911 | 4826064 |