Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019644.3 | NP_062618.2 | 6, 7 | 979 | ||
GenBank mRNA | AK302407.1 | - | 6, 7 | 933 | ||
BC007020.2 | - | 4, 5 | 513 | |||
BC032799.1 | - | 6, 7 | 740 | 5170634 | ||
D78334.1 | - | 6, 7 | 735 |