Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054027.4 | NP_473368.1 | 12 | 1938 | ||
XM_017009644.1 | XP_016865133.1 | 12 | 1538 | |||
GenBank mRNA | AB046801.1 | - | 12 | 1768 | ||
AK001799.1 | - | 8 | 1063 | |||
AK130676.1 | - | 7 | 902 | |||
AY358503.1 | - | 12 | 1689 | |||
BC014526.2 | - | 12 | 1663 | 3837372 |