Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033656.1 | NP_001028828.1 | 13 | 3669 | ||
XM_006256497.1 | XP_006256559.1 | 13 | 3541 | |||
GenBank mRNA | BC090336.1 | - | 5 | 1846 |